Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1600002H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028056.1 | NP_082332.1 | 4 | 515 | ||
GenBank mRNA | AK005395.1 | - | 3 | 715 | ||
AK146539.1 | - | 4 | 509 | |||
BC063767.1 | - | 3 | 433 | |||
BC072612.1 | - | 4 | 515 | 6414405 |