Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028059.2 | NP_082335.1 | 1 | 227 | ||
GenBank mRNA | AK005508.1 | - | 1 | 280 | ||
BC068018.1 | - | 1 | 227 | 6336013 |