Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF361644.1 | - | 3, 4 | 468 | ||
AK005532.1 | - | 3, 4 | 451 | |||
AK031742.1 | - | 3, 4 | 306 | |||
AK049358.1 | - | 3, 4 | 454 | |||
AK082340.1 | - | 1, 2 | 234 | |||
AK082749.1 | - | 1, 2 | 70 | |||
AK085617.1 | - | 3, 4 | 461 | |||
AK086940.1 | - | 3, 4 | 401 | |||
AK143428.1 | - | 3, 4 | 444 | |||
AK220209.1 | - | 3, 4 | 278 | |||
BB857917.1 | - | 3, 4 | 471 | |||
BC013549.1 | - | 3, 4 | 470 | 4222133 |