Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081229.1 | NP_001074698.1 | 1 | 1540 | ||
GenBank mRNA | AK132430.1 | - | 1 | 1353 | ||
BC117998.1 | - | 2 | 1341 | 40095084 |