Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 3 | 816 | ||
NM_001252579.1 | NP_001239508.1 | 3 | 662 | |||
NM_028072.5 | NP_082348.2 | 3 | 658 | |||
GenBank mRNA | AK028874.1 | - | 3 | 573 | ||
AK034712.1 | - | 3 | 643 | |||
AK036685.1 | - | 3 | 594 | |||
AK129316.1 | - | 3 | 816 | |||
AK133336.1 | - | 632 | ||||
AK165183.1 | - | 3 | 570 | |||
AY101177.1 | - | 2 | 477 | |||
BC062900.1 | - | 3 | 615 | |||
BC141086.1 | - | 2 | 411 | 9055698 | ||
CF736722.1 | - | 3 | 658 |