Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 3, 4 | 849 | ||
NM_001252579.1 | NP_001239508.1 | 3, 4 | 695 | |||
NM_028072.5 | NP_082348.2 | 3, 4 | 691 | |||
GenBank mRNA | AK028874.1 | - | 3, 4 | 606 | ||
AK034712.1 | - | 3, 4 | 676 | |||
AK036685.1 | - | 3, 4 | 627 | |||
AK081643.1 | - | 3, 4 | 649 | |||
AK129316.1 | - | 3, 4 | 849 | |||
AK133336.1 | - | 665 | ||||
AK165183.1 | - | 3, 4 | 603 | |||
AY101177.1 | - | 2, 3 | 510 | |||
BC062900.1 | - | 3, 4 | 648 | |||
BC141086.1 | - | 2, 3 | 444 | 9055698 | ||
CF736722.1 | - | 3, 4 | 691 |