Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028085.2 | NP_082361.2 | 2 | 649 | ||
GenBank mRNA | AF524030.1 | - | 2 | 606 | ||
AK008229.1 | - | 2 | 631 | |||
BC115784.1 | - | 2 | 610 | 40104967 | ||
BC118013.1 | - | 2 | 610 | 40104966 |