Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028089.3 | NP_082365.1 | 9 | 1430 | ||
GenBank mRNA | AK008580.1 | - | 9 | 1430 | ||
AY206875.1 | - | 9 | 1403 | |||
BC010824.1 | - | 9 | 1418 | 4219121 |