Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277296.1 | NP_001264225.1 | 11 | 1722 | ||
NM_001277297.1 | NP_001264226.1 | 11 | 1534 | |||
NM_028105.4 | NP_082381.1 | 11 | 1643 | |||
GenBank mRNA | AK011310.1 | - | 11 | 1494 | ||
AK050868.1 | - | 11 | 1624 | |||
AK145987.1 | - | 8 | 1202 | |||
AK154839.1 | - | 11 | 1589 | |||
BC010539.1 | - | 11 | 1536 | 4162365 |