Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028108.3 | NP_082384.1 | 5 | 658 | ||
GenBank mRNA | AK011160.1 | - | 5 | 584 | ||
AK135658.1 | - | 5 | 601 | |||
AK145345.1 | - | 5 | 586 | |||
AK156791.1 | - | 5 | 604 | |||
AK167141.1 | - | 6 | 778 | |||
AK168394.1 | - | 5 | 629 | |||
BC004086.1 | - | 5 | 570 | |||
BC023275.1 | - | 3 | 117 | |||
BC046283.1 | - | 4 | 458 | 6438976 | ||
BC057117.1 | - | 5 | 617 | 6409354 |