Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027656.2 | NP_081932.1 | 1 | 781 | ||
NM_028125.3 | NP_082401.1 | 2 | 952 | |||
GenBank mRNA | AK011461.1 | - | 2 | 953 | ||
AK016700.1 | - | 1 | 146 | |||
AK085310.1 | - | 2 | 954 | |||
AK155148.1 | - | 2 | 930 | |||
BC058179.1 | - | 2 | 928 | 5326477 |