Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027656.2 | NP_081932.1 | 1 | 326 | ||
NM_028125.3 | NP_082401.1 | 2 | 497 | |||
GenBank mRNA | AK011461.1 | - | 2 | 498 | ||
AK085310.1 | - | 2 | 498 | |||
AK155148.1 | - | 2 | 475 | |||
BC058179.1 | - | 2 | 473 | 5326477 |