Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028128.1 | NP_082404.1 | 10 | 943 | ||
GenBank mRNA | AK011489.1 | - | 10 | 954 | ||
AK150036.1 | - | 956 | ||||
AK170211.1 | - | 10 | 961 | |||
AK171372.1 | - | 1365 | ||||
BC023674.1 | - | 9 | 881 | |||
BC089001.1 | - | 10 | 943 | 6415420 |