Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028136.2 | NP_082412.2 | 25 | 2943 | ||
GenBank mRNA | AF448804.1 | - | 25 | 2943 | ||
AK013031.1 | - | 18 | 1935 | |||
BC033407.1 | - | 11 | 1245 | |||
BC138061.1 | - | 25 | 2919 | 8861080 |