Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028136.2 | NP_082412.2 | 22 | 2575 | ||
GenBank mRNA | AF448804.1 | - | 22 | 2575 | ||
AK013031.1 | - | 15 | 1567 | |||
BC033407.1 | - | 8 | 877 | |||
BC138061.1 | - | 22 | 2551 | 8861080 |