Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028140.4 | NP_082416.2 | 4 | 390 | ||
GenBank mRNA | AK013144.1 | - | 4 | 282 | ||
AK032080.1 | - | 4 | 411 | |||
AK035291.1 | - | 4 | 353 | |||
AK077957.1 | - | 4 | 391 | |||
BC010483.1 | - | 4 | 239 | |||
BC113183.1 | - | 4 | 321 | 40061891 | ||
BC113790.1 | - | 4 | 321 | 40061889 |