Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026998.3 | NP_081274.2 | 10 | 887 | ||
GenBank mRNA | AK008135.1 | - | 5 | 352 | ||
AK013158.1 | - | 10 | 888 | |||
AK030764.1 | - | 4 | 217 | |||
BC061028.1 | - | 11 | 928 | 6594585 |