Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2510009E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001881.2 | NP_001001881.1 | 2 | 661 | ||
GenBank mRNA | AK044207.1 | - | 2 | 579 | ||
BC052490.1 | - | 2 | 616 | |||
BC072629.1 | - | 2 | 610 | 6833156 |