Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028148.2 | NP_082424.2 | 8 | 806 | ||
GenBank mRNA | AK129478.1 | - | 8 | 698 | ||
BC058682.1 | - | 8 | 787 |