Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152812.3 | NP_690025.2 | 6 | 997 | ||
GenBank mRNA | AK011203.1 | - | 7 | 928 | ||
AK144761.1 | - | 6 | 820 | |||
AK155761.1 | - | 6 | 848 | |||
AK166445.1 | - | 6 | 827 | |||
AK166546.1 | - | 6 | 825 | |||
AK169134.1 | - | 6 | 812 | |||
AK172229.1 | - | 6 | 802 | |||
AK172338.1 | - | 6 | 802 | |||
BC031474.1 | - | 6 | 803 | 2599116 | ||
BC087552.1 | - | 6 | 794 | 6331952 |