Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028935.1 | NP_083211.1 | 8 | 1081 | ||
GenBank mRNA | AK005691.1 | - | 2 | 306 | ||
AK016584.1 | - | 10 | 2735 | |||
BC048749.1 | - | 7 | 789 |