Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285991.1 | NP_001272920.1 | 21 | 2360 | ||
NM_001285993.1 | NP_001272922.1 | 20 | 2261 | |||
NM_028162.4 | NP_082438.3 | 20 | 2294 | |||
GenBank mRNA | AK154975.1 | - | 21 | 2320 | ||
AK163366.1 | - | 20 | 2324 | |||
AK220251.1 | - | 20 | 2261 | |||
BC043113.1 | - | 20 | 2110 | 6407234 | ||
BC098328.1 | - | 15 | 1640 | 30357143 | ||
BU614210.1 | - | 2 | 38 |