Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285991.1 | NP_001272920.1 | 13 | 1312 | ||
NM_001285993.1 | NP_001272922.1 | 13 | 1279 | |||
NM_028162.4 | NP_082438.3 | 13 | 1312 | |||
GenBank mRNA | AK005461.1 | - | 13 | 1296 | ||
AK033710.1 | - | 13 | 1204 | |||
AK144467.1 | - | 13 | 1313 | |||
AK154975.1 | - | 13 | 1272 | |||
AK220251.1 | - | 13 | 1279 | |||
BC043113.1 | - | 13 | 1128 | 6407234 |