Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027403.4 | NP_081679.2 | 2 | 318 | ||
GenBank mRNA | AK005492.1 | - | 2 | 316 | ||
AK005540.1 | - | 2 | 313 | |||
AK005545.1 | - | 2 | 316 | |||
AK131670.1 | - | 2 | 316 | |||
BC100324.1 | - | 2 | 295 | 30715195 | ||
BC100325.1 | - | 2 | 295 | 30787148 | ||
BY033222.1 | - | 2 | 322 |