Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007839.1 | - | 1 | 39 | ||
AK028304.1 | - | 8 | 836 | |||
AK088814.1 | - | 8 | 826 | |||
AK142658.1 | - | 8 | 826 | |||
AK145291.1 | - | 8 | 821 | |||
AK150777.1 | - | 8 | 790 | |||
AK151240.1 | - | 8 | 789 | |||
AK151664.1 | - | 8 | 832 | |||
AK152338.1 | - | 8 | 832 | |||
AK152343.1 | - | 8 | 791 | |||
AK152991.1 | - | 8 | 791 | |||
AK155905.1 | - | 8 | 826 | |||
AK159203.1 | - | 8 | 826 | |||
AK159350.1 | - | 8 | 826 | |||
AK159878.1 | - | 8 | 835 | |||
AK168708.1 | - | 8 | 768 | |||
AY029764.1 | - | 8 | 718 | |||
BC012401.1 | - | 8 | 815 | 3967323 | ||
CT010238.1 | - | 8 | 683 |