Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810041L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163145.1 | NP_001156617.1 | 3 | 247 | ||
GenBank mRNA | AK129415.1 | - | 4 | 470 | ||
BC062953.1 | - | 3 | 247 | 6856297 | ||
BC119234.1 | - | 3 | 209 | 8733983 | ||
BC120551.1 | - | 3 | 209 | 40129474 |