Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011707.1 | NP_001011707.1 | 7 | 1169 | ||
GenBank mRNA | AK008512.2 | - | 6 | 995 | ||
AY227734.1 | - | 7 | 1169 | |||
BC147819.1 | - | 7 | 1166 | 9087536 | ||
BC147824.1 | - | 7 | 1166 | 9087541 |
Gene Symbol: Cyp2c65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028191.2 | NP_082467.2 | 7 | 1162 | ||
GenBank mRNA | AK008688.1 | - | 7 | 1092 | ||
AY227733.1 | - | 7 | 1153 | |||
BC022634.1 | - | 3 | 444 | |||
BC051089.1 | - | 7 | 1146 | |||
BC117531.1 | - | 7 | 1145 | 40106344 | ||
BC118514.1 | - | 7 | 1146 | 40106342 |