Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028195.3 | NP_082471.2 | 12 | 1202 | ||
GenBank mRNA | AK010908.1 | - | 2 | 135 | ||
AK152306.1 | - | 12 | 1108 | |||
AK152909.1 | - | 12 | 1108 | |||
AK157026.1 | - | 12 | 1108 | |||
AK220198.1 | - | 12 | 1131 | |||
BC139054.1 | - | 12 | 1175 | 8862076 | ||
BC145528.1 | - | 12 | 1202 |