Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028202.3 | NP_082478.1 | 3, 4 | 1597 | ||
GenBank mRNA | AK009491.1 | - | 3, 4 | 1513 | ||
AK078669.1 | - | 3, 4 | 1511 | |||
AK160799.1 | - | 3, 4 | 1457 |