Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081390.1 | NP_001074859.1 | 17 | 3228 | ||
NM_001293772.1 | NP_001280701.1 | 19 | 3958 | |||
NM_001293773.1 | NP_001280702.1 | 17 | 3038 | |||
NM_001293774.1 | NP_001280703.1 | 9 | 1804 | |||
GenBank mRNA | AF205078.1 | - | 7 | 982 | ||
AK010350.1 | - | 5 | 691 | |||
AK031696.1 | - | 17 | 3031 | |||
AK052489.1 | - | 19 | 3941 | |||
AK076423.1 | - | 16 | 2676 | |||
AK087573.1 | - | 9 | 1180 | |||
AK149663.1 | - | 9 | 1781 | |||
AK173081.1 | - | 17 | 2978 | |||
BC027364.1 | - | 17 | 2987 | |||
BC076588.1 | - | 10 | 2066 | |||
BC127081.1 | - | 10 | 1812 | |||
JX477684.1 | - | 17 | 3030 |