Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081381.1 | NP_001074850.1 | 3 | 294 | ||
GenBank mRNA | AK011270.1 | - | 4 | 450 | ||
AK075986.1 | - | 3 | 291 | |||
BC048732.1 | - | 3 | 269 | 6512688 | ||
CJ073544.1 | - | 3 | 294 |