Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrpprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028233.2 | NP_082509.2 | 2 | 548 | ||
GenBank mRNA | AK039751.1 | - | 2 | 324 | ||
AK138527.1 | - | 2 | 552 | |||
BC059862.1 | - | 2 | 282 | 6417407 | ||
BC060199.1 | - | 2 | 320 |