Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028235.1 | NP_082511.1 | 1 | 1249 | ||
GenBank mRNA | AK011097.1 | - | 1 | 1226 | ||
AK147134.1 | - | 1 | 1199 | |||
BC026507.1 | - | 1 | 441 | |||
BC151060.1 | - | 1 | 1249 | 9087973 | ||
BC151184.1 | - | 1 | 1249 | 9088073 | ||
BC157929.1 | - | 1 | 1318 | 9088102 |