Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011109.1 | - | 2 | 344 | ||
AK011167.1 | - | 2 | 447 | |||
AK020031.1 | - | 2 | 502 | |||
AK029454.1 | - | 2 | 335 | |||
AK146167.1 | - | 2 | 224 | |||
AK156717.1 | - | 3 | 767 | |||
AK168313.1 | - | 696 | ||||
BC027368.1 | - | 2 | 651 | |||
BE655013.1 | - | 2 | 253 |
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051339.1 | - | 3 | 1241 |