Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 6 | 1170 | ||
GenBank mRNA | AK011892.1 | - | 2 | 102 | ||
AK046669.1 | - | 6 | 1136 | |||
AK144950.1 | - | 2243 | ||||
AK164809.1 | - | 5 | 1412 | |||
AK166977.1 | - | 4 | 2511 | |||
AK173311.1 | - | 5 | 1418 | |||
BC031392.1 | - | 6 | 1114 | 4952407 |