Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030203.2 | NP_084479.1 | 1 | 404 | ||
GenBank mRNA | AK020999.1 | - | 1 | 315 | ||
AK030740.1 | - | 1 | 349 | |||
AK034070.1 | - | 1 | 371 | |||
AK034414.1 | - | 1 | 345 | |||
AK158197.1 | - | 1 | 372 | |||
BC017540.1 | - | 1 | 353 | 4511620 | ||
BC034656.1 | - | 1 | 352 | 5361000 | ||
BY248195.1 | - | 1 | 405 |