Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwf19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081077.1 | NP_001074546.1 | 14 | 1581 | ||
GenBank mRNA | AK087867.1 | - | 8 | 854 | ||
AK088543.1 | - | 16 | 2823 | |||
AK139654.1 | - | 14 | 1582 | |||
BC037640.1 | - | 14 | 1559 | 5096937 |