Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289591.1 | NP_001276520.1 | 7 | 1246 | ||
NM_001289592.1 | NP_001276521.1 | 8 | 1213 | |||
NM_028261.1 | NP_082537.1 | 8 | 1336 | |||
GenBank mRNA | AK012006.1 | - | 8 | 1283 | ||
AK077788.1 | - | 8 | 1237 | |||
AK089405.1 | - | 8 | 1314 | |||
AK146284.1 | - | 8 | 1209 | |||
AK153868.1 | - | 8 | 1207 | |||
AK157370.1 | - | 7 | 1148 | |||
AK158458.1 | - | 8 | 1122 | |||
AK170724.1 | - | 8 | 1314 | |||
AK171065.1 | - | 8 | 1300 | |||
AK171203.1 | - | 8 | 1300 | |||
AK171612.1 | - | 8 | 1267 | |||
AK172314.1 | - | 7 | 1079 | |||
AX775911.1 | - | 8 | 1223 | |||
BC027757.1 | - | 6 | 980 | |||
BC046640.1 | - | 7 | 1171 | 5067343 | ||
DQ910493.1 | - | 8 | 1134 | |||
FJ222242.1 | - | 6 | 998 |