Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163538.1 | NP_001157010.1 | 5 | 767 | ||
NM_028273.3 | NP_082549.2 | 5 | 764 | |||
GenBank mRNA | AK012159.1 | - | 5 | 743 | ||
AK034588.1 | - | 5 | 748 | |||
AK049246.1 | - | 5 | 727 | |||
AK146216.1 | - | 5 | 768 | |||
AK169643.1 | - | 5 | 730 | |||
AK187787.1 | - | 1 | 104 | |||
BC021317.1 | - | 5 | 717 | |||
BC052179.1 | - | 5 | 691 | |||
BC138924.1 | - | 5 | 747 | 8861946 | ||
BC138925.1 | - | 5 | 747 | 8861947 | ||
BC145467.1 | - | 5 | 750 | 9054029 |