Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130172.1 | NP_001123644.1 | 12 | 1670 | ||
NM_001130173.1 | NP_001123645.1 | 13 | 2042 | |||
NM_001161656.1 | NP_001155128.1 | 13 | 2033 | |||
NM_001161657.1 | NP_001155129.1 | 11 | 1424 | |||
NM_001161658.1 | NP_001155130.1 | 13 | 1994 | |||
NM_001161659.1 | NP_001155131.1 | 12 | 1679 | |||
NM_001161660.1 | NP_001155132.1 | 11 | 1574 | |||
NM_005375.3 | NP_005366.2 | 12 | 1679 | |||
NR_134958.1 | 13 | 1801 | ||||
NR_134959.1 | 13 | 1792 | ||||
NR_134960.1 | 13 | 1735 | ||||
NR_134961.1 | 13 | 1862 | ||||
NR_134962.1 | 12 | 1580 | ||||
NR_134963.1 | 12 | 1679 | ||||
NR_134964.1 | 13 | 1726 | ||||
NR_134965.1 | 14 | 1857 | ||||
GenBank mRNA | AF104863.1 | - | 12 | 1557 | ||
AJ606317.1 | - | 13 | 1801 | |||
AJ606318.1 | - | 13 | 1862 | |||
AJ606319.1 | - | 13 | 2042 | |||
AJ606320.1 | - | 12 | 1670 | |||
AJ606321.1 | - | 13 | 1792 | |||
AJ606322.1 | - | 13 | 1735 | |||
AJ606323.1 | - | 12 | 1679 | |||
AJ606324.1 | - | 12 | 1679 | |||
AJ616235.1 | - | 12 | 1594 | |||
AJ616802.1 | - | 1 | 19 | |||
AY787443.1 | - | 12 | 1480 | |||
AY787444.1 | - | 10 | 1187 | |||
AY787445.1 | - | 12 | 1530 | |||
AY787446.1 | - | 12 | 1381 | |||
AY787447.1 | - | 11 | 1375 | |||
AY787448.1 | - | 11 | 1225 | |||
AY787449.1 | - | 12 | 1281 | |||
AY787450.1 | - | 13 | 1536 | |||
AY787451.1 | - | 12 | 1480 | |||
AY787452.1 | - | 12 | 1480 | |||
AY787453.1 | - | 12 | 1480 | |||
AY787454.1 | - | 13 | 1602 | |||
AY787455.1 | - | 14 | 1974 | |||
AY787456.1 | - | 14 | 1658 | |||
AY787457.1 | - | 14 | 1573 | |||
AY787458.1 | - | 13 | 1527 | |||
AY787459.1 | - | 13 | 1593 | |||
AY787460.1 | - | 14 | 1956 | |||
AY787461.1 | - | 13 | 1834 | |||
AY787462.1 | - | 13 | 1983 | |||
AY787463.1 | - | 13 | 1749 | |||
AY787464.1 | - | 13 | 1663 | |||
AY787465.1 | - | 13 | 2077 | |||
AY787466.1 | - | 14 | 1719 | |||
AY787467.1 | - | 13 | 1843 | |||
AY787468.1 | - | 13 | 1795 | |||
AY787469.1 | - | 14 | 1851 | |||
AY787470.1 | - | 12 | 1395 | |||
AY787471.1 | - | 13 | 1451 | |||
AY787472.1 | - | 13 | 1578 | |||
AY787473.1 | - | 13 | 1992 | |||
AY787474.1 | - | 13 | 1758 | |||
AY787475.1 | - | 12 | 1480 | |||
BC035996.1 | - | 6 | 710 | |||
BC064955.1 | - | 12 | 1631 | 6069320 | ||
M13665.1 | - | 10 | 1342 | |||
M15024.1 | - | 12 | 1593 | |||
X52125.1 | - | 12 | 1668 | |||
X52126.1 | - | 12 | 1596 |