Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028281.1 | NP_082557.1 | 2 | 228 | ||
GenBank mRNA | AA013784.1 | - | 2 | 224 | ||
AK012465.1 | - | 2 | 226 | |||
BC028642.1 | - | 2 | 184 | 1194896 | ||
BY157602.1 | - | 2 | 229 |