Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025392.1 | NP_001020563.1 | 5 | 1803 | ||
NM_001025393.1 | NP_001020564.1 | 5 | 1803 | |||
NM_153787.2 | NP_722482.1 | 5 | 1797 | |||
GenBank mRNA | AK012290.1 | - | 1 | 465 | ||
AK079110.1 | - | 1 | 53 | |||
AK141834.1 | - | 5 | 1750 | |||
AK149724.1 | - | 5 | 1756 | |||
AK150017.1 | - | 5 | 1756 | |||
AK161608.1 | - | 693 | ||||
AK161692.1 | - | 5 | 1801 | |||
BC029682.1 | - | 5 | 1734 | |||
BC034300.1 | - | 5 | 1729 | 4507876 | ||
BC086624.1 | - | 5 | 1785 |