Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028296.3 | NP_082572.1 | 3 | 1162 | ||
GenBank mRNA | AB080741.1 | - | 4 | 1118 | ||
AK012302.1 | - | 4 | 1125 | |||
AK035758.1 | - | 3 | 397 | |||
AK038695.1 | - | 4 | 1042 | |||
AK132222.1 | - | 3 | 1003 | |||
AK162576.1 | - | 3 | 280 | |||
BC017606.1 | - | 3 | 354 | 4507552 | ||
BC066111.1 | - | 4 | 1118 | 30360743 |