Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028303.3 | NP_082579.1 | 5 | 422 | ||
GenBank mRNA | AK007454.1 | - | 5 | 376 | ||
AK012624.1 | - | 5 | 423 | |||
AK032327.1 | - | 59 | ||||
BC004608.1 | - | 5 | 380 | 3585336 | ||
BC022181.1 | - | 4 | 885 |