Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178625.3 | NP_848740.3 | 12 | 1395 | ||
GenBank mRNA | AK029366.1 | - | 9 | 1084 | ||
AK044877.1 | - | 12 | 1411 | |||
AK078544.1 | - | 12 | 1409 | |||
AK089486.1 | - | 1270 | ||||
AK089962.1 | - | 12 | 1410 | |||
AK170465.1 | - | 1270 | ||||
BC052187.1 | - | 12 | 1438 | 6393580 | ||
BC057109.1 | - | 12 | 1395 | 6400080 |