Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278610.1 | NP_001265539.1 | 5 | 724 | ||
NM_002466.3 | NP_002457.1 | 6 | 796 | |||
GenBank mRNA | AK223482.1 | - | 6 | 531 | ||
AK303249.1 | - | 5 | 629 | |||
AK314791.1 | - | 6 | 701 | |||
BC007585.1 | - | 6 | 721 | 3162656 | ||
BC053555.1 | - | 6 | 690 | 6140320 | ||
X13293.1 | - | 6 | 658 |