Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159595.1 | NP_001153067.1 | 9 | 1233 | ||
NM_178112.5 | NP_835213.3 | 9 | 1233 | |||
GenBank mRNA | AK012736.1 | - | 9 | 1095 | ||
AK036255.1 | - | 1 | 425 | |||
AK049513.1 | - | 9 | 1204 | |||
AK140829.1 | - | 9 | 1228 | |||
AK169552.1 | - | 9 | 1202 | |||
BC049092.1 | - | 9 | 1163 | 6402150 | ||
BC057618.1 | - | 9 | 1163 | 6402150 | ||
BC060149.1 | - | 1 | 15 |