Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700094K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033166.2 | NP_001028338.1 | 1, 2 | 226 | ||
NM_001037279.1 | NP_001032356.1 | 1, 2 | 226 | |||
GenBank mRNA | AK012609.1 | - | 1, 2 | 204 | ||
AK142622.1 | - | 1, 2 | 216 | |||
AK167638.1 | - | 1, 2 | 198 | |||
AK171972.1 | - | 1, 2 | 217 | |||
BC056177.1 | - | 1, 2 | 219 | 5683334 | ||
BY239096.1 | - | 1, 2 | 288 | |||
CA318611.1 | - | 1, 2 | 123 |