Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700094K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033166.2 | NP_001028338.1 | 3 | 390 | ||
NM_001037279.1 | NP_001032356.1 | 3 | 390 | |||
GenBank mRNA | AK012609.1 | - | 3 | 368 | ||
AK142622.1 | - | 3 | 380 | |||
AK167638.1 | - | 3 | 362 | |||
AK171972.1 | - | 3 | 381 | |||
BC056177.1 | - | 3 | 383 | 5683334 | ||
CA318611.1 | - | 3 | 287 |