Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160326.1 | NP_001153798.1 | 1 | 224 | ||
GenBank mRNA | AK012850.1 | - | 1 | 240 | ||
AY425619.1 | - | 1 | 209 | |||
BC100483.1 | - | 2 | 179 | |||
BC115492.1 | - | 2 | 209 | 40100869 | ||
BC115493.1 | - | 2 | 209 | |||
BU057067.1 | - | 1 | 224 | |||
CB057846.1 | - | 2 | 165 |